Penicillin G Natriumsalz 98%

Lieferant: Thermo Fisher Scientific
Gefahr

455690250 455690010 455690050
ACRO455690250EA 97.4 EUR
ACRO455690250 ACRO455690010 ACRO455690050
Penicillin G Natriumsalz 98%
Penicillin G Natriumsalz
Formel: C₁₆H₁₇N₂NaO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00069666
CAS-Nummer: 69-57-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
pH 5.0 to 7.5
Specific optical rotation +293° to +303° (20°C, 589 nm) (c=2, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR