2-Pyridinsulfonsäure 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Pyridine-2-sulphonic acid
455760050
ACRO455760050EA
82.4
EUR
ACRO455760050
2-Pyridinsulfonsäure 97%
2-Pyridinesulphonic acid
Formel:
C₅H₅NO₃S Molecular Weight: 159,1631 g/mol Schmelzpunkt: 244…249 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234080 CAS-Nummer: 15103-48-7 UN: 2585 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to tan Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 245 to 252 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...