4-Aminopyridine-2-carboxylic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Aminopyridine-2-carboxylic Acid
455772500
ACRO455772500EA
114
EUR
ACRO455772500
4-Aminopyridine-2-carboxylic acid 97%
4-Aminopyridine-2-carboxylic acid
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: 319 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02685671 CAS-Nummer: 100047-36-7 |
Spezifikation Testergebnisse
Appearance | Beige Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...