7-Azaindole-5-boronic acid pinacol ester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
7-Azaindole-5-boronic Acid Pinacol Ester
455850010
ACRO455850010EA
190
EUR
ACRO455850010
7-Azaindole-5-boronic acid pinacol ester 97%
7-Azaindole-5-boronic acid pinacol ester
Formel:
C₁₃H₁₇BN₂O₂ Schmelzpunkt: 243…245,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08060937 CAS-Nummer: 754214-56-7 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Melting point | 243 to 249 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...