1-Boc-piperidin-4-ylboronsäurepinakolester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: tert-Butyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)piperidine-1-carboxylate, Piperidine-4-boronic acid, pinacol ester, N-BOC protected

456050050
ACRO456050050EA 360 EUR
ACRO456050050
1-Boc-piperidin-4-ylboronsäurepinakolester 97%
1-Boc-piperidin-4-ylboronsäurepinakolester
Formel: C₁₆H₃₀BNO₄
Molecular Weight: 311,2247 g/mol
Schmelzpunkt: 56,5…57 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09997792
CAS-Nummer: 1048970-17-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 60 to 65 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR