Oxaliplatin 99%

Lieferant: Thermo Fisher Scientific
Gefahr

456131000
ACRO456131000EA 140 EUR
ACRO456131000
Oxaliplatin 99%
Oxaliplatin
Formel: C₈H₁₄N₂O₄Pt
Lagertemperatur: Gefrierschrank
MDL: MFCD00866327
CAS-Nummer: 61825-94-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (1 g, 105°C, 2 h)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR