Pemetrexed disodium salt 2.5 hydrate 98%

Lieferant: Thermo Scientific
Gefahr

456180010
ACRO456180010EA 157 EUR
ACRO456180010
Pemetrexed disodium salt 2.5 hydrate 98%
Pemetrexed disodium salt 2.5 hydrate
Formel: C₂₀H₁₉N₅Na₂O₆·⁵/₂H₂O
Molecular Weight: 516,42 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13194950
CAS-Nummer: 357166-30-4

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Water ≤10 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR