Capecitabin 99%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 5'-Deoxy-5-fluoro-N4-[(pentyloxy)carbonyl]cytidine, Pentyl (1-((2R,3R,4S,5R)-3,4-dihydroxy-5-methyltetrahydrofuran-2-yl)-5-fluoro-2-oxo-1,2-dihydropyrimidin-4-yl)carbamate

456241000
ACRO456241000EA 226 EUR
ACRO456241000
Capecitabin 99%
Capecitabine
Formel: C₁₅H₂₂FN₃O₆
Molecular Weight: 359,35 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00930626
CAS-Nummer: 154361-50-9

Bestellen

Spezifikation Testergebnisse

Appearance White to almost white Powder
Infrared spectrum Conforms
Water ≤0.3 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR