Paliperidone 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-9-hydroxy-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one, 9-Hydroxyrisperidone

456250010
ACRO456250010EA 406 EUR
ACRO456250010
Paliperidone 98%
Paliperidone
Formel: C₂₃H₂₇FN₄O₃
Molecular Weight: 426,49 g/mol
Schmelzpunkt: 171 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00871802
CAS-Nummer: 144598-75-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Crystalline powder
Infrared spectrum Conforms
Melting point 168 to 175 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR