1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: DABSO, 1,4-Diazabicyclo[2.2.2]octane-1,4-diium-1,4-disulphinate, DABCO-Bis(sulphur dioxide), Bis(sulphur dioxide)-1,4-diazabicyclo[2.2.2]octane

456350010 456350050
ACRO456350010EA 45.8 EUR
ACRO456350010 ACRO456350050
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct 97%
1,4-Diazabicyclo[2.2.2]octanebis(sulphur dioxide)adduct
Formel: C₆H₁₂N₂O₄S₂
Molecular Weight: 240.29 g/mol
Schmelzpunkt: 168…172 °C
Lagertemperatur: Kühlschrank
MDL: MFCD22377802
CAS-Nummer: 119752-83-9
UN: 1325
ADR: 4.1 II

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 168 to 172 °C
Assay ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR