4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 4-(5-(p-Tolyl)-3-(trifluoromethyl)-1H-pyrazol-1-yl)benzenesulphonamide, Celecoxib

456451000
ACRO456451000EA 197 EUR
ACRO456451000
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide 98%
4-[5-(4-Methylphenyl)-3-(trifluoromethyl)pyrazol-1-yl]benzenesulphonamide
Formel: C₁₇H₁₄F₃N₃O₂S
Molecular Weight: 381,38 g/mol
Schmelzpunkt: 156…157 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00941298
CAS-Nummer: 169590-42-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 159 to 165 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR