Tetrahydropyran-4-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

456720050
ACRO456720050EA 35.5 EUR
ACRO456720050
Tetrahydropyran-4-carbonsäure 97%
Tetrahydropyran-4-carbonsäure
Formel: C₆H₁₀O₃
Siedepunkt: 115 °C (20 mmHg)
Schmelzpunkt: 87…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031016
CAS-Nummer: 5337-03-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 87 to 91 °C
Water ≤1.5 %
Titration with NaOH 96.5 to 103.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR