Ciprofloxacin hydrochloride hydrate 98%
Lieferant: Thermo Fisher Scientific
456880050
456880250
ACRO456880050EA
33.5
EUR
ACRO456880050
ACRO456880250
Ciprofloxacin hydrochloride hydrate 98%
Ciprofloxacin hydrochloride hydrate
Formel:
C₁₇H₁₈FN₃O₃·HCl·xH₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00242856 CAS-Nummer: 128074-72-6 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Water | 4.0 to 7.0 % |
HPLC | ≥97.5 % |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...