Gemcitabine 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Amino-1-((2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one
456890010
456890050
ACRO456890010EA
98.2
EUR
ACRO456890010
ACRO456890050
Gemcitabine 98%
Gemcitabine
Formel:
C₉H₁₁F₂N₃O₄ Molecular Weight: 263.2 g/mol Schmelzpunkt: 292 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00869720 CAS-Nummer: 95058-81-4 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...