(S)-Tetrahydrofuran-3-amine hydrochloride 95%

Lieferant: Thermo Fisher Scientific
Achtung

456930050
ACRO456930050EA 412 EUR
ACRO456930050
(S)-Tetrahydrofuran-3-amine hydrochloride 95%
(S)-Tetrahydrofuran-3-amine hydrochloride
Formel: C₄H₁₀ClNO
Schmelzpunkt: 140…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445642
CAS-Nummer: 204512-95-8

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Powder or solid
Infrared spectrum Conforms
Titration Argentometric ≥94.0 to ≤106.0 %
Specific optical rotation -2° to -8° (20°C, 589 nm) c=0.5 in Methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR