Decyldecanoat 98%

Lieferant: Thermo Fisher Scientific

457110250 457115000
ACRO457110250EA 37.5 EUR
ACRO457110250 ACRO457115000
Decyldecanoat 98%
Decyldecanoat
Formel: C₂₀H₄₀O₂
Siedepunkt: 174 °C (2 mmHg)
Dichte: 0,859 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00059280
CAS-Nummer: 1654-86-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4410 to 1.4430 (20°C, 589 nm)
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR