4'-Benzyloxy-2'-hydroxyacetophenon 98%

Lieferant: Thermo Fisher Scientific

457130010
ACRO457130010EA 110 EUR
ACRO457130010
4'-Benzyloxy-2'-hydroxyacetophenon 98%
4'-Benzyloxy-2'-hydroxyacetophenon
Formel: C₁₅H₁₄O₃
Schmelzpunkt: 104…106 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00054612
CAS-Nummer: 29682-12-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow to light brown Powder
Infrared spectrum Conforms
Melting point 101 to 107 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR