3-Amino-6-brom-2-pyrazincarbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

457420050
ACRO457420050EA 317 EUR
ACRO457420050
3-Amino-6-brom-2-pyrazincarbonsäure 97%
3-Amino-6-bromopyrazine-2-carboxylic acid
Formel: C₅H₄BrN₃O₂
Molecular Weight: 218.01 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275681
CAS-Nummer: 486424-37-7

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR