1,6-Naphthyridin 96%

Lieferant: Thermo Scientific

Synonyme: Pyrido[4,3-b]pyridine, 1,6-Diazanaphthalene

457670010
ACRO457670010EA 293 EUR
ACRO457670010
1,6-Naphthyridin 96%
1,6-Naphthyridine
Formel: C₈H₆N₂
Molecular Weight: 130,15 g/mol
Schmelzpunkt: 36…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00059750
CAS-Nummer: 253-72-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to brown Low melting solid
Infrared spectrum Conforms
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR