3-Aminoisonicotinsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Aminoisonicotinic Acid, 3-Aminopyridine-4-carboxylic acid
457690050
ACRO457690050EA
245
EUR
ACRO457690050
3-Aminoisonicotinsäure 98%
3-Aminoisonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: 275…276 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137840 CAS-Nummer: 7579-20-6 |
Spezifikation Testergebnisse
Appearance | Yellow to light brown Powder |
Infrared spectrum | Conforms |
Melting point | ≥270 °C |
HPLC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...