Moxifloxacin hydrochloride
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-Cyclopropyl-6-fluoro-7-((4aS,7aS)-hexahydro-1H-pyrrolo[3,4-b]pyridin-6(2H)-yl)-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Moxifloxacin HCl
457960010
ACRO457960010EA
215
EUR
ACRO457960010
Moxifloxacin hydrochloride
Moxifloxacin hydrochloride
Formel:
C₂₁H₂₅ClFN₃O₄ Molecular Weight: 437,896 g/mol Schmelzpunkt: 324…325 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00949117 CAS-Nummer: 186826-86-8 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow Powder |
Infrared spectrum | Conforms |
Water | ≤5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...