Abacavir sulphate 99%

Lieferant: Thermo Fisher Scientific
Gefahr

458860010 458860050
ACRO458860010EA 55 EUR
ACRO458860010 ACRO458860050
Abacavir sulphate 99%
Abacavir sulphate
Formel: C₂₈H₃₈N₁₂O₆S
Lagertemperatur: Kühlschrank
MDL: MFCD04112763
CAS-Nummer: 188062-50-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR