Theophyllin-7-essigsäure 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(1,3-Dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-7H-purin-7-yl)acetic acid
459251000
ACRO459251000EA
58.5
EUR
ACRO459251000
Theophyllin-7-essigsäure 98%
Theophyllin-7-essigsäure
Formel:
C₉H₁₀N₄O₄ Molecular Weight: 238,20 g/mol Schmelzpunkt: 270 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00022832 CAS-Nummer: 652-37-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 268 to 272 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...