2-Amino-6-(trifluoromethoxy)benzothiazole 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
459360010
459360050
459360100
ACRO459360010EA
60.9
EUR
ACRO459360010
ACRO459360050
ACRO459360100
2-Amino-6-(trifluoromethoxy)benzothiazole 98%
2-Amino-6-(trifluoromethoxy)benzothiazole
Formel:
C₈H₅F₃N₂OS Schmelzpunkt: 114…118 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00210213 CAS-Nummer: 1744-22-5 UN: 2811 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 116 to 120 °C |
Water | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...