Robenidine hydrochloride 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3-Bis[(4-chlorobenzylidene)amino]guanidine hydrochloride

459400250
ACRO459400250EA 185 EUR
ACRO459400250
Robenidine hydrochloride 98%
Robenidine hydrochloride
Formel: C₁₅H₁₃Cl₂N₅·ClH
Molecular Weight: 370,67 g/mol
Schmelzpunkt: 252…254 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD01724404
CAS-Nummer: 25875-50-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR