Cefadroxil 95-105 %
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Cefadroxil monohydrate, (6R,7R)-7-((R)-2-Amino-2-(4-hydroxyphenyl)acetamido)-3-methyl-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
459470010
ACRO459470010EA
0
EUR
ACRO459470010
Cefadroxil 95-105 %
Cefadroxil
Formel:
C₁₆H₁₇N₃O₅S Molecular Weight: 363,388 g/mol (anhy) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00865091 CAS-Nummer: 66592-87-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | 4.2 to 6.0 % |
HPLC | ≥95 % |
pH | 4.0 to 6.0 |
Specific optical rotation | 165° to 178° (20°C, 589 nm) (1 % in water DS) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...