Chloramphenicol sodium succinate 98-102%

Lieferant: Thermo Fisher Scientific
Achtung

459530050 459530250
ACRO459530050EA 46.7 EUR
ACRO459530050 ACRO459530250
Chloramphenicol sodium succinate 98-102%
Chloramphenicol sodium succinate
Formel: C₁₅H₁₅Cl₂N₂NaO₈
Molecular Weight: 445.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00083598
CAS-Nummer: 982-57-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Assay 98.0 to 102.0 % on dried substance
Water ≤2 %
Specific optical rotation +5.0° to +8.0° (20°C, 589 nm) (5 % in water) (on dried substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR