Roxithromycin 96%

Lieferant: Thermo Scientific
Achtung

Synonyme: (3R,4S,5S,6R,7R,9R,11S,12R,13S,14R,E)-6-(((2S,3R,4S,6R)-4-(Dimethylamino)-3-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-14-ethyl-7,12,13-trihydroxy-4-(((2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyltetrahydro-2H-pyran-2-yl)oxy)-10-(((2-methoxyethoxy)methoxy)imino)-3,5,7,9,11,13-hexamethyloxacyclotetradecan-2-one

459570050
ACRO459570050EA 80 EUR
ACRO459570050
Roxithromycin 96%
Roxithromycin

White to off-white Powder

Formel: C₄₁H₇₆N₂O₁₅
Molecular Weight: 837,04 g/mol
Schmelzpunkt: 111…118 °C
Flammpunkt: >110 °C
MDL: MFCD00214389
CAS-Nummer: 80214-83-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤3 %
HPLC ≥95.0 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR