Heptadecafluoroctansulfonamid 95%

Lieferant: Thermo Fisher Scientific
Achtung

459640010 459640050
ACRO459640010EA 93.2 EUR
ACRO459640010 ACRO459640050
Heptadecafluoroctansulfonamid 95%
Heptadecafluoroctansulfonamid
Formel: C₈H₂F₁₇NO₂S
Schmelzpunkt: 154…155 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094345
CAS-Nummer: 754-91-6
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Solid
Infrared spectrum Conforms
GC ≥94 % (mix of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR