4-Chloro-1H-imidazole 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
5-Chloro-1H-imidazole
460030010
ACRO460030010EA
171
EUR
ACRO460030010
4-Chloro-1H-imidazole 98%
4-Chloro-1H-imidazole
Formel:
C₃H₃ClN₂ Schmelzpunkt: 118…120 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09033087 CAS-Nummer: 15965-31-8 |
Spezifikation Testergebnisse
Appearance | White to off-white Solid |
Infrared spectrum | Conforms |
Melting point | 118 to 121 °C |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...