7-Chinolinamin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: Quinolin-7-amine

460050010
ACRO460050010EA 133 EUR
ACRO460050010
7-Chinolinamin 97%
7-Aminoquinoline
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Schmelzpunkt: 80…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646243
CAS-Nummer: 580-19-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to orange Powder
Infrared spectrum Conforms
Melting point 85 to 90 °C
Water ≤1 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR