Efaproxiral sodium 98%

Lieferant: Thermo Fisher Scientific
Achtung

460100010 460102500
ACRO460100010EA 322 EUR
ACRO460100010 ACRO460102500
Efaproxiral sodium 98%
Efaproxiral sodium
Formel: C₂₀H₂₂NNaO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD02093864
CAS-Nummer: 170787-99-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤13 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR