Fluorometholon 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 9-Fluoro-11,17-dihydroxy-6-methylpregna-1,4-diene-3,20-dione

460200010
ACRO460200010EA 109 EUR
ACRO460200010
Fluorometholon 97%
Fluorometholone
Formel: C₂₂H₂₉FO₄
Molecular Weight: 376,47 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00056461
CAS-Nummer: 426-13-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
Solubility (2.5 % in pyridine) clear colorless to light yellow
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR