Methyl nervonate (15 cis), Nervonic acid methyl ester 99% analytischer Standard für GC

Lieferant: Thermo Fisher Scientific

Synonyme: Methyl 15(Z)-tetracosenoate, 15(Z)-Tetracosenoic acid methyl ester, Methyl nervonate (15 cis), Nervonic acid methyl ester

460371000 460375000
ACRO460371000EA 70.6 EUR
ACRO460371000 ACRO460375000
Methyl nervonate (15 cis), Nervonic acid methyl ester 99% analytischer Standard für GC
Methyl nervonate
Formel: C₂₅H₄₈O₂
Molecular Weight: 380.65 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00056324
CAS-Nummer: 2733-88-2

Bestellen

Spezifikation Testergebnisse

Appearance White low melting solid
Infrared spectrum Conforms
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR