Moxalactam sodium salt

Lieferant: Thermo Fisher Scientific
Gefahr

460410010
ACRO460410010EA 210 EUR
ACRO460410010
Moxalactam sodium salt
Moxalactam sodium salt
Formel: C₂₀H₁₈N₆Na₂O₉S
Lagertemperatur: Kühlschrank
MDL: MFCD03427565
CAS-Nummer: 64953-12-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥88 (mixture of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR