β-Pseudouridine 98%
Lieferant: Thermo Fisher Scientific
460451000
ACRO460451000EA
294
EUR
ACRO460451000
β-Pseudouridine 98%
β-Pseudouridine
Formel:
C₉H₁₂N₂O₆ Schmelzpunkt: 230 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00038458 CAS-Nummer: 1445-07-4 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 212 to 217 °C |
Loss on drying | ≤2 % |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...