Droperidol

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-[1-[3-(p-Fluorobenzoyl)propyl]-1,2,3,6-tetrahydro-4-pyridyl]-2-benzimidazolinone, 1-(1-(4-(4-Fluorophenyl)-4-oxobutyl)-1,2,3,6-tetrahydropyridin-4-yl)-1H-benzo[d]imidazol-2(3H)-one

460840010
ACRO460840010EA 131 EUR
ACRO460840010
Droperidol
Droperidol
Formel: C₂₂H₂₂FN₃O₂
Molecular Weight: 379,43 g/mol
Schmelzpunkt: 148…149 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00083290
CAS-Nummer: 548-73-2

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤5 % (70°C, 4 h) (vaccuum)
Titration with HClO4 98 to 102 % (on dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR