Lamotrigin 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, 3,5-Diamino-6-(2,3-dichlorophenyl)-1,2,4-triazine

460850010 460850050
ACRO460850010EA 54.2 EUR
ACRO460850010 ACRO460850050
Lamotrigin 98%
Lamotrigine
Formel: C₉H₇Cl₂N₅
Molecular Weight: 256.09 g/mol
Schmelzpunkt: 218 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00865333
CAS-Nummer: 84057-84-1
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 216 to 220 °C
Loss on drying ≤0.5 % (105 °C, 3 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR