Oxyclozanide 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2,3,5-Trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide
460881000
ACRO460881000EA
79.5
EUR
ACRO460881000
Oxyclozanide 98%
Oxyclozanide
Formel:
C₁₃H₆Cl₅NO₃ Molecular Weight: 401,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00864507 CAS-Nummer: 2277-92-1 UN: 3077 ADR: 9,III |
Spezifikation Testergebnisse
Appearance | Cream Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...