1,2-Indanedione 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
460920010
ACRO460920010EA
103
EUR
ACRO460920010
1,2-Indanedione 95%
1,2-Indanedione
Formel:
C₉H₆O₂ Schmelzpunkt: 117…124 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463378 CAS-Nummer: 16214-27-0 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 110 to 120 °C |
Water | ≤2.5 % |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...