Atazanavir
Lieferant: Thermo Fisher Scientific
Synonyme:
1,14-Dimethyl (3S,8S,9S,12S)-3,12-bis(1,1-dimethylethyl)-8-hydroxy-4,11-dioxo-9-(phenylmethyl)-6-[[4-(2-pyridinyl)phenyl]methyl]-2,5,6,10,13-pentaazatetradecanedioate
460981000
ACRO460981000EA
187
EUR
ACRO460981000
Atazanavir
Atazanavir
Formel:
C₃₈H₅₂N₆O₇ Schmelzpunkt: 207…209 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD08435966 CAS-Nummer: 198904-31-3 |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 205 to 209 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...