Bicalutamid 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: N-(4-Cyano-3-(trifluoromethyl)phenyl)-3-((4-fluorophenyl)sulphonyl)-2-hydroxy-2-methylpropanamide, N-[4-Cyano-3-(trifluoromethyl)phenyl]-3-[(4-fluorophenyl)sulphonyl]-2-hydroxy-2-methylpropionamide

461000010
ACRO461000010EA 167 EUR
ACRO461000010
Bicalutamid 97%
Bicalutamide
Formel: C₁₈H₁₄F₄N₂O₄S
Molecular Weight: 430,37 g/mol
Schmelzpunkt: 196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00869971
CAS-Nummer: 90357-06-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 194 to 198 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 4 h)
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR