Cyclizine hydrochloride 98%

Lieferant: Thermo Fisher Scientific
Gefahr

461040010
ACRO461040010EA 184 EUR
ACRO461040010
Cyclizine hydrochloride 98%
Cyclizine hydrochloride
Formel: C₁₈H₂₂N₂·HCl
Schmelzpunkt: 291…293 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00067334
CAS-Nummer: 303-25-3
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (120°C, 3 h)
Titration with HClO4 ≥97.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR