Lamivudin 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (-)-1-[(2R,5S)-2-(Hydroxymethyl)-1,3-oxathiolan-5-yl]cytosine, 4-Amino-1-((2R,5S)-2-(hydroxymethyl)-1,3-oxathiolan-5-yl)pyrimidin-2(1H)-one, 3TC

461090010
ACRO461090010EA 128 EUR
ACRO461090010
Lamivudin 98%
Lamivudine
Formel: C₈H₁₁N₃O₃S
Molecular Weight: 229,2600 g/mol
Schmelzpunkt: 160…162 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00869739
CAS-Nummer: 134678-17-4

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 175 to 179 °C
Water ≤0.5 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR