Medroxyprogesterone acetate 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 17alpha-Hydroxy-6alpha-methyl-4-pregnene-3,20-dione 17-Acetate, Medroxyprogesterone 17-Acetate, 17alpha-Acetoxy-6alpha-methylprogesterone

461120010 461120050
ACRO461120010EA 55.7 EUR
ACRO461120010 ACRO461120050
Medroxyprogesterone acetate 97%
Medroxyprogesterone acetate
Formel: C₂₄H₃₄O₄
Schmelzpunkt: 206…208 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00010483
CAS-Nummer: 71-58-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 203 to 209 °C
Loss on drying ≤1 %
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR