Mefloquin -hydrochlorid

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (S)-[2,8-Bis(trifluoromethyl)quinolin-4-yl][(2R)-piperidin-2-yl]methanol hydrochloride, rel-(S)-(2,8-Bis(trifluoromethyl)quinolin-4-yl)((R)-piperidin-2-yl)methanol hydrochloride, Mefloquine HCI

461130010
ACRO461130010EA 108 EUR
ACRO461130010
Mefloquin -hydrochlorid
Mefloquin -hydrochlorid
Formel: C₁₇H₁₆F₆N₂O
Schmelzpunkt: 253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00797519
CAS-Nummer: 51773-92-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Titration Argentometric ≥97.5 % (on dry substance)
Water ≤3 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR