Pyridostigmine bromide 98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 3-((Dimethylcarbamoyl)oxy)-1-methylpyridin-1-ium bromide, 3-(Dimethylcarbamoyloxy)-1-methylpyridinium Bromide

461190010 461190050 461190250
ACRO461190010EA 56.1 EUR
ACRO461190010 ACRO461190050 ACRO461190250
Pyridostigmine bromide 98%
Pyridostigmine bromide
Formel: C₉H₁₃BrN₂O₂
Schmelzpunkt: 154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079283
CAS-Nummer: 101-26-8
UN: 2811
ADR: 6.1,II

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 153 to 157 °C
Loss on drying ≤2 % (1 g, 105 °C)
Titration with HClO4 97.5 to 102.5 % (on dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR