Saquinavir mesylate 98%
Lieferant: Thermo Scientific
Synonyme:
(S)-N1-((2S,3R)-4-((3S,4AS,8aS)-3-(tert-butylcarbamoyl)octahydroisoquinolin-2(1H)-yl)-3-hydroxy-1-phenylbutan-2-yl)-2-(quinoline-2-carboxamido)succinamide methanesulfonate
461231000
ACRO461231000EA
106
EUR
ACRO461231000
Saquinavir mesylate 98%
Saquinavir mesylate
Formel:
C₃₈H₅₀N₆O₅·CH₄O₃S Molecular Weight: 766,95 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00944907 CAS-Nummer: 149845-06-7 |
Spezifikation Testergebnisse
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...