Tenofovirdisoproxilfumarat 98%

Lieferant: Thermo Fisher Scientific

461250010 461250050 461250250
ACRO461250010EA 35.3 EUR
ACRO461250010 ACRO461250050 ACRO461250250
Tenofovirdisoproxilfumarat 98%
Tenofovir disoproxil fumarate
Formel: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
Lagertemperatur: Kühlschrank
MDL: MFCD08141829
CAS-Nummer: 202138-50-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %
Fumaric acid 17.5 to 19


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR