Valganciclovir hydrochloride 97+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-[(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)methoxy]-3-hydroxypropyl (2S)-2-amino-3-methylbutanoate hydrochloride hydrate, Valcyte, Valganciclovir HCl, (2S)-2-((2-Amino-6-oxo-1H-purin-9(6H)-yl)methoxy)-3-hydroxypropyl 2-amino-3-methylbutanoate hydrochloride

461270010
ACRO461270010EA 369 EUR
ACRO461270010
Valganciclovir hydrochloride 97+%
Valganciclovir hydrochloride
Formel: C₁₄H₂₂N₆O₅·HCl
Molecular Weight: 390.83 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08460118
CAS-Nummer: 175865-59-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤8 %
HPLC ≥97.0 % ( sum of two diastereomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR